Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003941.2 | NP_001003941.1 | 3, 4 | 571 | ||
NM_002541.3 | NP_002532.2 | 3, 4 | 571 | |||
XM_005249759.4 | XP_005249816.1 | 3, 4 | 503 | |||
XM_011515408.2 | XP_011513710.1 | 4, 5 | 590 | |||
GenBank mRNA | AK304690.1 | - | 2, 3 | 309 | ||
AK314179.1 | - | 3, 4 | 493 | |||
AU140451.1 | - | 3, 4 | 525 | |||
BC004964.1 | - | 2, 3 | 454 | 3528538 | ||
BC009580.1 | - | 3, 4 | 505 | |||
BC014617.2 | - | 2, 3 | 454 | 3010095 | ||
BC128550.1 | - | 1, 2 | 85 | |||
D10523.1 | - | 3, 4 | 469 | |||
DQ925674.1 | - | 1, 2 | 27 |