Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017372.2 | NP_001017372.1 | 3 | 1716 | ||
NM_001317984.1 | NP_001304913.1 | 4 | 1540 | |||
NM_014031.4 | NP_054750.1 | 4 | 1491 | |||
GenBank mRNA | AF064254.1 | - | 3 | 858 | ||
AK313456.1 | - | 4 | 1139 | |||
AL833117.1 | - | 3 | 1289 | |||
BC041945.1 | - | 3 | 1715 | 5301306 | ||
BX537383.1 | - | 4 | 1101 | |||
CR457129.1 | - | 3 | 705 |