Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145939.1 | NP_001139411.1 | 5 | 719 | ||
NM_001145940.1 | NP_001139412.1 | 5 | 480 | |||
NM_002573.3 | NP_002564.1 | 4 | 697 | |||
GenBank mRNA | BC003016.1 | - | 4 | 624 | 2822228 | |
BC007863.2 | - | 4 | 495 | 4301185 | ||
CR407626.1 | - | 4 | 362 | |||
D63391.1 | - | 4 | 475 |