Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017421.3 | NP_059117.3 | 5, 6 | 765 | ||
XM_005267018.4 | XP_005267075.1 | 4, 5 | 538 | |||
XM_006715500.3 | XP_006715563.1 | 6, 7 | 904 | |||
XM_011535898.2 | XP_011534200.1 | 5, 6 | 662 | |||
XM_011535899.1 | XP_011534201.1 | 4, 5 | 593 | |||
XM_017010933.1 | XP_016866422.1 | 6, 7 | 762 | |||
GenBank mRNA | AF193016.1 | - | 5, 6 | 702 | ||
AF351615.1 | - | 5, 6 | 770 | |||
AK056955.1 | - | 5, 6 | 763 | |||
AL136726.1 | - | 5, 6 | 763 | |||
BC001534.1 | - | 5, 6 | 736 | |||
BC015634.2 | - | 5, 6 | 726 | |||
BC063463.1 | - | 5, 6 | 741 | 5196692 |