Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018484.3 | NP_060954.1 | 8 | 1738 | ||
XM_011545167.1 | XP_011543469.1 | 7 | 1388 | |||
GenBank mRNA | AB026116.1 | - | 8 | 1391 | ||
AK075224.1 | - | 8 | 1084 | |||
AK222869.1 | - | 8 | 1396 | |||
AK290791.1 | - | 8 | 1721 | |||
BC034384.1 | - | 8 | 1713 | 5190509 |