Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019844.3 | NP_062818.1 | 6 | 689 | ||
GenBank mRNA | AF187815.1 | - | 4 | 448 | ||
AJ251506.1 | - | 4 | 492 | |||
AK055874.1 | - | 6 | 667 | |||
AY342017.1 | - | 4 | 462 | |||
BC022843.1 | - | 5 | 573 | |||
BC058006.1 | - | 5 | 578 | |||
BC070264.1 | - | 5 | 552 | |||
BC105597.1 | - | 5 | 578 | |||
BG567966.1 | - | 5 | 577 |