Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001290.1 | NP_001001290.1 | 13 | 1720 | ||
NM_020041.2 | NP_064425.2 | 12 | 1709 | |||
XM_017008460.1 | XP_016863949.1 | 11 | 3366 | |||
GenBank mRNA | AF210317.1 | - | 12 | 1702 | ||
BC018897.2 | - | 13 | 1720 | 3949549 | ||
BC110414.1 | - | 12 | 1739 | 5762360 |