Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020309.3 | NP_064705.1 | 7, 8 | 1046 | ||
GenBank mRNA | AB032436.1 | - | 7, 8 | 895 | ||
AK289622.1 | - | 4, 5 | 454 | |||
AK294226.1 | - | 7, 8 | 852 | |||
AK294405.1 | - | 7, 8 | 895 | |||
BC059379.1 | - | 7, 8 | 1021 | 5262415 |