Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021196.3 | NP_067019.3 | 14 | 2020 | ||
NM_133478.2 | NP_597812.1 | 19 | 2416 | |||
GenBank mRNA | AB209752.1 | - | 15 | 2823 | ||
AF243499.2 | - | 14 | 2018 | |||
AF293337.2 | - | 18 | 2365 | |||
AF293338.2 | - | 14 | 2018 | |||
AF452248.1 | - | 16 | 2199 | |||
AF453528.1 | - | 16 | 2208 | |||
BC109221.2 | - | 14 | 1875 | 40008543 |