Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135146.1 | NP_001128618.1 | 9 | 2235 | ||
NM_001135148.1 | NP_001128620.1 | 8 | 1672 | |||
NM_022154.5 | NP_071437.3 | 8 | 2357 | |||
XM_005263177.1 | XP_005263234.1 | 9 | 2144 | |||
XM_011532182.1 | XP_011530484.1 | 7 | 2170 | |||
XM_017008540.1 | XP_016864029.1 | 9 | 2152 | |||
XM_017008541.1 | XP_016864030.1 | 11 | 2529 | |||
GenBank mRNA | AB040120.1 | - | 8 | 2305 | ||
AK027652.1 | - | 8 | 1672 | |||
AL049963.2 | - | 1 | 333 | |||
BC001320.1 | - | 2 | 739 | |||
BC012125.1 | - | 9 | 2211 | 4650026 |