Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040454.1 | NP_001035544.1 | 4 | 697 | ||
NM_022911.2 | NP_075062.2 | 5 | 609 | |||
NM_134263.2 | NP_599025.2 | 5 | 609 | |||
NM_134426.2 | NP_602298.2 | 5 | 609 | |||
GenBank mRNA | AB033288.1 | - | 5 | 572 | ||
AB102713.1 | - | 5 | 654 | |||
AB208936.1 | - | 3 | 1755 | |||
AF279265.1 | - | 4 | 697 | |||
AF288410.1 | - | 5 | 557 | |||
AF416721.1 | - | 5 | 557 | |||
BC017697.1 | - | 5 | 558 | 4398446 |