Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193476.1 | NP_001180405.1 | 14 | 1723 | ||
NM_052961.3 | NP_443193.1 | 14 | 1747 | |||
NM_138718.2 | NP_619732.2 | 12 | 1470 | |||
XM_011514294.2 | XP_011512596.1 | 13 | 1888 | |||
XM_017010235.1 | XP_016865724.1 | 14 | 1773 | |||
GenBank mRNA | AF314959.1 | - | 13 | 1617 | ||
AF331522.1 | - | 14 | 1691 | |||
AK055314.1 | - | 4 | 1193 | |||
AK057276.1 | - | 4 | 266 | |||
AK097410.1 | - | 8 | 809 | |||
AK122928.1 | - | 14 | 1723 | |||
AK125189.1 | - | 13 | 2047 | |||
BC025408.2 | - | 12 | 1463 | 4823098 |