Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080282.3 | NP_525021.3 | 14 | 2238 | ||
GenBank mRNA | AL832004.2 | - | 14 | 1944 | ||
AY028900.1 | - | 14 | 2238 | |||
AY247065.1 | - | 14 | 2269 | |||
BC136460.1 | - | 11 | 1443 | |||
BC144124.1 | - | 11 | 1512 |