Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270880.1 | NP_001257809.1 | 2 | 289 | ||
NM_138335.2 | NP_612208.1 | 2 | 289 | |||
GenBank mRNA | AB209904.1 | - | 2 | 200 | ||
AF247786.1 | - | 2 | 250 | |||
AK055639.1 | - | 2 | 209 | |||
AY173948.1 | - | 2 | 226 | |||
BC015532.2 | - | 1 | 120 | 3914747 | ||
BP274548.1 | - | 2 | 289 |