Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130058.1 | NP_001123530.1 | 8 | 571 | ||
NM_001320283.1 | NP_001307212.1 | 6 | 532 | |||
NM_001320285.1 | NP_001307214.1 | 10 | 731 | |||
NM_001320287.1 | NP_001307216.1 | 9 | 664 | |||
NM_152697.4 | NP_689910.2 | 8 | 571 | |||
XM_006710445.3 | XP_006710508.1 | 6 | 458 | |||
XM_011540986.2 | XP_011539288.1 | 7 | 549 | |||
XM_011540987.2 | XP_011539289.1 | 9 | 626 | |||
XM_017000607.1 | XP_016856096.1 | 7 | 597 | |||
XM_017000608.1 | XP_016856097.1 | 7 | 596 | |||
XM_017000609.1 | XP_016856098.1 | 10 | 1915 | |||
XM_017000610.1 | XP_016856099.1 | 10 | 993 | |||
XM_017000611.1 | XP_016856100.1 | 8 | 678 | |||
GenBank mRNA | AK093170.1 | - | 8 | 573 | ||
AK128063.1 | - | 9 | 624 | |||
AK296924.1 | - | 7 | 610 | |||
AK296941.1 | - | 6 | 431 | |||
AK299573.1 | - | 6 | 532 | |||
BC028743.2 | - | 8 | 544 | 4828797 | ||
BC034580.1 | - | 10 | 710 | 4824861 | ||
BC051740.1 | - | 10 | 710 | 4824908 | ||
DC374918.1 | - | 6 | 532 |