Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206974.1 | NP_001193903.1 | 3, 4 | 463 | ||
NM_001930.3 | NP_001921.1 | 3, 4 | 612 | |||
NM_013406.2 | NP_037538.1 | 3, 4 | 612 | |||
NR_038192.1 | 3, 4 | 612 | ||||
XM_011527770.1 | XP_011526072.1 | 3, 4 | 612 | |||
XM_011527771.2 | XP_011526073.1 | 3, 4 | 623 | |||
GenBank mRNA | AK291553.1 | - | 3, 4 | 593 | ||
AK304780.1 | - | 3, 4 | 558 | |||
AL520040.3 | - | 3, 4 | 388 | |||
AY336090.1 | - | 3, 4 | 501 | |||
BC000333.2 | - | 3, 4 | 559 | 2822554 | ||
BC014016.2 | - | 3, 4 | 574 | 4129051 | ||
BU844677.1 | - | 3, 4 | 591 | |||
L39068.1 | - | 3, 4 | 528 | |||
U26266.1 | - | 3, 4 | 493 | |||
U32178.1 | - | 3, 4 | 493 | |||
U40579.1 | - | 3, 4 | 574 | |||
U79262.1 | - | 3, 4 | 586 |