Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004446.2 | NP_004437.2 | 7 | 953 | ||
GenBank mRNA | AK296755.1 | - | 7 | 825 | ||
AK307534.1 | - | 5 | 527 | |||
AY493416.1 | - | 5 | 500 | |||
BC015494.1 | - | 7 | 741 | |||
BC034797.1 | - | 7 | 749 | |||
BC046156.1 | - | 7 | 953 | |||
BC058921.1 | - | 7 | 790 | |||
BC067841.1 | - | 6 | 730 | |||
BC105983.1 | - | 7 | 751 | |||
BC126275.1 | - | 7 | 821 | 8991991 | ||
BC136465.1 | - | 7 | 721 | 9020454 | ||
CR933648.1 | - | 7 | 792 | |||
JQ080075.1 | - | 7 | 683 | |||
X07466.1 | - | 3 | 183 | |||
X54326.1 | - | 5 | 525 |