Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164716.1 | NP_001158188.1 | 1 | 930 | ||
GenBank mRNA | AF066859.1 | - | 1 | 113 | ||
AK056607.1 | - | 1 | 189 | |||
AK299461.1 | - | 1 | 189 | |||
AK314474.1 | - | 1 | 117 | |||
BC029305.1 | - | 1 | 162 | |||
BC126392.1 | - | 1 | 512 | 8992108 | ||
BC130514.1 | - | 1 | 512 | 40146545 |