Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145933.1 | NP_001139405.1 | 13 | 2114 | ||
NM_001145934.1 | NP_001139406.1 | 12 | 2059 | |||
NM_012253.3 | NP_036385.3 | 13 | 2132 | |||
GenBank mRNA | AK292261.1 | - | 13 | 2008 | ||
AK301724.1 | - | 13 | 1933 | |||
AK302044.1 | - | 13 | 1990 | |||
BC025382.2 | - | 13 | 2008 | 4825931 | ||
X91817.1 | - | 12 | 1898 |