Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRMT2B SIRNA Assays ›
Gene Symbol:
TRMT2B
Gene Name:
tRNA methyltransferase 2 homolog B
Gene Aliases:
CXorf34, dJ341D10.3
Chromosome Location:
Chr.X: 100973358 - 101052118 on Build GRCh38
UniGene ID:
Hs.496501
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118030
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRMT2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167970.1
NP_001161442.1
14
2043
NM_001167971.1
NP_001161443.1
14
2024
NM_001167972.1
NP_001161444.1
13
1986
NM_024917.5
NP_079193.2
14
2436
XM_005262195.2
XP_005262252.1
15
2159
XM_005262196.2
XP_005262253.1
13
1954
XM_005262201.2
XP_005262258.1
11
1534
XM_006724705.2
XP_006724768.1
14
2098
XM_011531042.2
XP_011529344.1
11
1652
XM_011531044.2
XP_011529346.1
13
1713
XM_017029858.1
XP_016885347.1
15
2544
XM_017029859.1
XP_016885348.1
14
1859
XM_017029860.1
XP_016885349.1
15
2158
XM_017029861.1
XP_016885350.1
14
2068
XM_017029862.1
XP_016885351.1
13
2296
XM_017029863.1
XP_016885352.1
15
1968
XM_017029864.1
XP_016885353.1
13
2160
XM_017029865.1
XP_016885354.1
13
1788
XM_017029866.1
XP_016885355.1
14
1851
XM_017029867.1
XP_016885356.1
14
1888
XM_017029868.1
XP_016885357.1
13
1761
XM_017029869.1
XP_016885358.1
12
2105
XM_017029870.1
XP_016885359.1
12
1695
XM_017029871.1
XP_016885360.1
12
1677
XM_017029872.1
XP_016885361.1
14
1901
XM_017029873.1
XP_016885362.1
14
1876
XM_017029874.1
XP_016885363.1
13
2218
XM_017029875.1
XP_016885364.1
12
1765
GenBank mRNA
AK022749.1
-
13
1799
AL832849.1
-
14
1962
BC007526.1
-
14
2055
2988022
BC008067.2
-
7
985
BC009437.1
-
13
1929
3504468
BC020116.1
-
14
1997
4342798
BC034272.2
-
14
2401
4933351
BG747665.1
-
3
422
BX647969.1
-
13
1983
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
RNA methylation
tRNA processing
protein binding
S-adenosylmethionine-dependent tRNA (m5U54) methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0