Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252675.1 | NP_001239604.1 | 4 | 730 | ||
NM_001252676.1 | NP_001239605.1 | 3 | 557 | |||
NM_001252677.1 | NP_001239606.1 | 4 | 730 | |||
NM_032501.3 | NP_115890.2 | 4 | 730 | |||
GenBank mRNA | AK092295.1 | - | 2 | 147 | ||
AK125058.1 | - | 2 | 1933 | |||
AK127566.1 | - | 4 | 676 | |||
AK296306.1 | - | 3 | 554 | |||
AK314440.1 | - | 4 | 653 | |||
BC039261.1 | - | 4 | 689 | 5298849 | ||
BC044588.1 | - | 4 | 655 | 5260496 | ||
BC055008.1 | - | 4 | 655 | 4838976 | ||
BC114612.1 | - | 4 | 521 |