Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252675.1 | NP_001239604.1 | 14 | 2904 | ||
NM_001252676.1 | NP_001239605.1 | 13 | 2737 | |||
NM_032501.3 | NP_115890.2 | 14 | 2910 | |||
XM_006723659.1 | XP_006723722.1 | 10 | 2256 | |||
XM_011529388.1 | XP_011527690.1 | 13 | 2695 | |||
GenBank mRNA | AB058749.1 | - | 9 | 1827 | ||
AK027817.1 | - | 8 | 1733 | |||
AK090851.1 | - | 2 | 1670 | |||
AK092295.1 | - | 12 | 2319 | |||
AK125058.1 | - | 12 | 4113 | |||
AK125923.1 | - | 2 | 3997 | |||
AL832939.1 | - | 10 | 1979 | |||
AL833041.1 | - | 2 | 1163 | |||
BC032076.1 | - | 7 | 1598 | |||
BC039261.1 | - | 14 | 2869 | 5298849 |