Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139169.4 | NP_631908.1 | 4 | 541 | ||
GenBank mRNA | AF448144.1 | - | 4 | 497 | ||
AK223567.1 | - | 4 | 486 | |||
AK302800.1 | - | 4 | 275 | |||
AK312805.1 | - | 4 | 507 | |||
BC030601.2 | - | 4 | 541 | 4822413 | ||
BX648479.1 | - | 4 | 505 | |||
BX648709.1 | - | 4 | 506 |