Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257994.1 | NP_001244923.1 | 3, 4 | 1102 | ||
NM_022727.5 | NP_073564.3 | 3, 4 | 1102 | |||
NM_182984.4 | NP_892029.2 | 4, 5 | 978 | |||
XM_011530139.2 | XP_011528441.1 | 4, 5 | 909 | |||
XM_017028770.1 | XP_016884259.1 | 3, 4 | 1112 | |||
GenBank mRNA | AK025106.1 | - | 2, 3 | 498 | ||
AK057029.1 | - | 3, 4 | 1052 | |||
BC013352.2 | - | 3, 4 | 747 | 4054100 | ||
BC017184.2 | - | 4, 5 | 864 | 3347641 | ||
BC108251.1 | - | 3, 4 | 782 | 3852106 | ||
CR456354.1 | - | 3, 4 | 823 |