Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242466.1 | NP_001229395.1 | 6 | 835 | ||
NM_181504.3 | NP_852556.2 | 7 | 972 | |||
NM_181523.2 | NP_852664.1 | 13 | 2272 | |||
NM_181524.1 | NP_852665.1 | 7 | 1104 | |||
XM_005248542.3 | XP_005248599.1 | 13 | 2295 | |||
XM_011543493.2 | XP_011541795.1 | 12 | 1925 | |||
XM_017009585.1 | XP_016865074.1 | 13 | 3064 | |||
XM_017009586.1 | XP_016865075.1 | 12 | 1885 | |||
GenBank mRNA | AF279367.1 | - | 7 | 1104 | ||
AK000121.1 | - | 7 | 1018 | |||
AK094785.1 | - | 6 | 835 | |||
AK126345.1 | - | 6 | 2235 | |||
AK223613.1 | - | 7 | 952 | |||
AK294919.1 | - | 5 | 1091 | |||
AK299743.1 | - | 11 | 1301 | |||
BC030815.1 | - | 7 | 944 | 4290954 | ||
BC094795.1 | - | 13 | 2252 | 30528412 | ||
JX133164.1 | - | 6 | 694 | |||
JX133165.1 | - | 6 | 694 | |||
M61906.1 | - | 12 | 1734 |