Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021939.3 | NP_068758.3 | 10 | 1970 | ||
XM_011525099.2 | XP_011523401.1 | 11 | 2475 | |||
XM_011525100.2 | XP_011523402.1 | 10 | 1602 | |||
GenBank mRNA | AB045981.1 | - | 10 | 1731 | ||
AF337909.1 | - | 10 | 1645 | |||
AK025694.1 | - | 9 | 1402 | |||
AK025874.1 | - | 7 | 1019 | |||
AK092042.1 | - | 9 | 1919 | |||
AK092708.1 | - | 9 | 1634 | |||
AK172811.1 | - | 10 | 1677 | |||
AK296107.1 | - | 10 | 1467 | |||
AL832985.1 | - | 10 | 1712 | |||
BC016467.1 | - | 10 | 1705 | 4154344 | ||
BX537452.1 | - | 10 | 1725 |