Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ABCC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032583.3 | NP_115972.2 | 7 | 857 | ||
NM_033151.3 | NP_149163.2 | 6 | 1143 | |||
NM_145186.2 | NP_660187.1 | 7 | 857 | |||
XM_011523397.1 | XP_011521699.1 | 3 | 363 | |||
XM_017023795.1 | XP_016879284.1 | 7 | 3471 | |||
XM_017023796.1 | XP_016879285.1 | 8 | 944 | |||
XM_017023797.1 | XP_016879286.1 | 8 | 1072 | |||
XM_017023798.1 | XP_016879287.1 | 7 | 898 | |||
XM_017023799.1 | XP_016879288.1 | 7 | 880 | |||
XM_017023800.1 | XP_016879289.1 | 7 | 871 | |||
XM_017023801.1 | XP_016879290.1 | 7 | 3471 | |||
XM_017023803.1 | XP_016879292.1 | 7 | 3471 | |||
GenBank mRNA | AF352582.1 | - | 7 | 892 | ||
AF367202.1 | - | 7 | 871 | |||
AF411579.1 | - | 7 | 871 | |||
AK301547.1 | - | 7 | 898 | |||
AY040219.1 | - | 6 | 1143 |