Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003400.3 | NP_003391.1 | 5, 6 | 1084 | ||
XM_005264546.1 | XP_005264603.1 | 3, 4 | 192 | |||
XM_006712094.2 | XP_006712157.1 | 6, 7 | 1227 | |||
XM_011533097.1 | XP_011531399.1 | 5, 6 | 393 | |||
XM_011533098.1 | XP_011531400.1 | 5, 6 | 1061 | |||
XM_011533099.2 | XP_011531401.1 | 5, 6 | 2034 | |||
GenBank mRNA | AK289920.1 | - | 5, 6 | 763 | ||
AK312426.1 | - | 5, 6 | 522 | |||
AL833550.1 | - | 6, 7 | 2023 | |||
BC032847.2 | - | 6, 7 | 485 | 5267242 | ||
BX647758.1 | - | 5, 6 | 1066 | |||
CR749840.1 | - | 6, 7 | 858 | |||
D89729.1 | - | 4, 5 | 374 | |||
Y08614.2 | - | 5, 6 | 394 |