Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025208.4 | NP_079484.1 | 2 | 648 | ||
NM_033135.3 | NP_149126.1 | 2 | 630 | |||
GenBank mRNA | AB033832.1 | - | 2 | 432 | ||
AF113216.1 | - | 2 | 382 | |||
AF335584.1 | - | 2 | 457 | |||
AF336376.1 | - | 2 | 451 | |||
AK292801.1 | - | 2 | 456 | |||
AY027517.1 | - | 2 | 368 | |||
AY027518.1 | - | 2 | 350 | |||
AY359116.1 | - | 2 | 289 | |||
BC030645.2 | - | 2 | 710 | 4824526 | ||
BG722948.1 | - | 2 | 520 |