Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11824
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
8, 9
1223
NM_001242353.1
NP_001229282.1
8, 9
1309
NM_001242354.1
NP_001229283.1
7, 8
995
NM_002540.4
NP_002531.3
10, 11
1511
NM_153432.1
NP_702910.1
7, 8
1069
NM_153433.1
NP_702911.1
8, 9
1236
NM_153435.1
NP_702913.1
8, 9
1129
NM_153436.1
NP_702914.1
7, 8
893
NM_153437.2
NP_702915.1
8, 9
1159
NM_153439.1
NP_702917.1
7, 8
1012
NM_153440.1
NP_702918.1
7, 8
939
XM_005252010.2
XP_005252067.1
7, 8
1021
XM_005252011.2
XP_005252068.1
7, 8
961
XM_005252012.3
XP_005252069.1
8, 9
1211
XM_005252013.3
XP_005252070.1
8, 9
1158
XM_005252014.3
XP_005252071.1
8, 9
1207
XM_005252017.4
XP_005252074.1
8, 9
1112
XM_005252018.2
XP_005252075.1
9, 10
1317
XM_005252019.3
XP_005252076.1
9, 10
1336
XM_005252022.2
XP_005252079.1
8, 9
1135
XM_005252023.2
XP_005252080.1
8, 9
1106
XM_005252024.3
XP_005252081.1
8, 9
1125
XM_005252026.4
XP_005252083.1
8, 9
1054
XM_005252029.2
XP_005252086.1
8, 9
1135
XM_006717126.3
XP_006717189.1
8, 9
1211
XM_011518722.2
XP_011517024.1
8, 9
1135
XM_011518723.2
XP_011517025.1
8, 9
1078
XM_011518724.2
XP_011517026.1
9, 10
1299
XM_011518725.2
XP_011517027.1
9, 10
1303
XM_011518726.2
XP_011517028.1
9, 10
1321
XM_011518728.2
XP_011517030.1
8, 9
1072
XM_011518729.2
XP_011517031.1
8, 9
1008
XM_011518730.2
XP_011517032.1
8, 9
1135
XM_011518731.2
XP_011517033.1
7, 8
964
XM_011518732.2
XP_011517034.1
8, 9
921
XM_011518733.2
XP_011517035.1
7, 8
907
XM_011518744.1
XP_011517046.1
7, 8
961
XM_017014764.1
XP_016870253.1
9, 10
1292
XM_017014765.1
XP_016870254.1
9, 10
1311
XM_017014766.1
XP_016870255.1
7, 8
1021
XM_017014767.1
XP_016870256.1
9, 10
1303
XM_017014768.1
XP_016870257.1
8, 9
1095
XM_017014769.1
XP_016870258.1
8, 9
1124
XM_017014770.1
XP_016870259.1
9, 10
1279
XM_017014771.1
XP_016870260.1
9, 10
1260
XM_017014772.1
XP_016870261.1
9, 10
1336
XM_017014773.1
XP_016870262.1
9, 10
1312
XM_017014774.1
XP_016870263.1
8, 9
1022
XM_017014775.1
XP_016870264.1
9, 10
1279
XM_017014776.1
XP_016870265.1
9, 10
1260
XM_017014777.1
XP_016870266.1
8, 9
1062
XM_017014778.1
XP_016870267.1
8, 9
1165
XM_017014779.1
XP_016870268.1
7, 8
906
XM_017014780.1
XP_016870269.1
8, 9
1089
XM_017014781.1
XP_016870270.1
7, 8
844
GenBank mRNA
AF012549.1
-
8, 9
1099
AF053970.1
-
6, 7
798
AK093367.1
-
7, 8
1019
AK295662.1
-
6, 7
739
AK299303.1
-
8, 9
1071
AK300793.1
-
7, 8
844
AK301754.1
-
7, 8
936
AK301842.1
-
7, 8
861
AK301855.1
-
7, 8
915
AK302684.1
-
8, 9
1163
AK308278.1
-
6, 7
775
AK309917.1
-
8, 9
1014
AY319414.1
-
7, 8
1045
AY366499.1
-
8, 9
1006
BC010629.1
-
10, 11
1309
3874501
BC091500.1
-
8, 9
1123
30378678
DQ444713.1
-
6, 7
836
DQ444714.1
-
6, 7
779
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0