Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005740.2 | NP_005731.1 | 4 | 1405 | ||
GenBank mRNA | AL035366.1 | - | 4 | 1398 | ||
BC002968.1 | - | 4 | 1372 | 3543962 |