Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018728.3 | NP_061198.2 | 41 | 5434 | ||
XM_011521781.2 | XP_011520083.1 | 41 | 5350 | |||
XM_017022408.1 | XP_016877897.1 | 40 | 5299 | |||
XM_017022409.1 | XP_016877898.1 | 36 | 4741 | |||
XM_017022410.1 | XP_016877899.1 | 40 | 5284 | |||
GenBank mRNA | AF272390.1 | - | 41 | 5291 | ||
BC144611.1 | - | 38 | 4890 |