Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308301.1 | NP_001295230.1 | 12 | 1737 | ||
NM_182493.2 | NP_872299.2 | 13 | 2556 | |||
XM_006721334.3 | XP_006721397.1 | 13 | 2024 | |||
GenBank mRNA | AB451455.1 | - | 13 | 2440 | ||
AJ247087.1 | - | 13 | 2556 | |||
AK299443.1 | - | 12 | 1737 | |||
BC109097.1 | - | 13 | 2453 | 40034724 |