Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004798.3 | NP_004789.1 | 4, 5 | 1801 | ||
GenBank mRNA | AB002357.1 | - | 4, 5 | 1788 | ||
AK098431.1 | - | 3, 4 | 346 | |||
AK299877.1 | - | 4, 5 | 724 | |||
BC136310.1 | - | 3, 4 | 1694 | 9020297 | ||
BC136311.1 | - | 3, 4 | 1694 | 9020298 |