Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015074.3 | NP_055889.2 | 3 | 392 | ||
NM_183416.3 | NP_904325.2 | 3 | 392 | |||
GenBank mRNA | AB011163.2 | - | 3 | 341 | ||
AB017133.1 | - | 2 | 224 | |||
AB040881.2 | - | 3 | 451 | |||
AB088210.1 | - | 2 | 140 | |||
AB088211.1 | - | 2 | 140 | |||
AB088212.1 | - | 2 | 140 | |||
AB088213.1 | - | 2 | 140 | |||
AF257176.1 | - | 3 | 293 | |||
AK308121.1 | - | 3 | 392 | |||
AY043362.1 | - | 2 | 144 | |||
AY139835.1 | - | 3 | 293 | |||
BC115395.2 | - | 2 | 228 | 40067993 | ||
BF940695.1 | - | 3 | 325 |