Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002597.4 | NP_002588.3 | 4 | 1056 | ||
NM_022576.3 | NP_072098.1 | 3 | 1018 | |||
XM_011509603.2 | XP_011507905.1 | 3 | 2074 | |||
GenBank mRNA | AF076464.1 | - | 3 | 1019 | ||
BM715757.1 | - | 1 | 632 | |||
M33478.1 | - | 4 | 1033 |