Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145754.3 | NP_665697.1 | 7 | 780 | ||
XM_005272357.3 | XP_005272414.1 | 7 | 870 | |||
XM_011517362.2 | XP_011515664.1 | 7 | 871 | |||
GenBank mRNA | AK055045.1 | - | 3 | 158 | ||
BC017311.2 | - | 7 | 762 | 5090622 | ||
BM751216.1 | - | 4 | 343 |