Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000096.3 | NP_000087.1 | 1, 2 | 407 | ||
NR_046371.1 | 1, 2 | 407 | ||||
XM_006713499.2 | XP_006713562.1 | 1, 2 | 412 | |||
XM_006713500.3 | XP_006713563.1 | 1, 2 | 412 | |||
XM_006713501.2 | XP_006713564.1 | 1, 2 | 412 | |||
XM_011512435.1 | XP_011510737.1 | 1, 2 | 412 | |||
XM_017005734.1 | XP_016861223.1 | 1, 2 | 412 | |||
XM_017005735.1 | XP_016861224.1 | 1, 2 | 412 | |||
GenBank mRNA | AK291219.1 | - | 1, 2 | 189 | ||
AK299272.1 | - | 1, 2 | 193 | |||
BC070304.1 | - | 1, 2 | 360 | |||
BC094709.1 | - | 1, 2 | 191 | |||
BC142714.1 | - | 1, 2 | 163 | 40148840 | ||
BC146663.1 | - | 1, 2 | 163 | 40148048 | ||
BC146801.1 | - | 1, 2 | 187 | |||
CA391602.1 | - | 1, 2 | 404 | |||
DC334592.1 | - | 1, 2 | 407 | |||
M13699.1 | - | 1, 2 | 154 |