Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118564
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
6
1450
NM_001098624.2
NP_001092094.1
6
1380
NM_001193277.1
NP_001180206.1
6
1239
NM_001193278.1
NP_001180207.1
6
1258
NM_001193279.1
NP_001180208.1
5
1105
NM_001193280.1
NP_001180209.1
5
991
NM_033289.1
NP_150631.1
6
1336
NM_033290.3
NP_150632.1
6
1242
XM_005274536.1
XP_005274593.1
7
1415
XM_005274537.1
XP_005274594.1
7
1392
XM_006724492.3
XP_006724555.1
7
2014
XM_011545525.2
XP_011543827.1
9
1716
XM_011545526.2
XP_011543828.1
9
1751
XM_011545527.2
XP_011543829.1
7
1661
XM_017029534.1
XP_016885023.1
7
1581
XM_017029535.1
XP_016885024.1
9
1602
XM_017029536.1
XP_016885025.1
8
1678
XM_017029537.1
XP_016885026.1
8
1449
XM_017029538.1
XP_016885027.1
6
1148
XM_017029539.1
XP_016885028.1
6
1310
XM_017029540.1
XP_016885029.1
8
1563
XM_017029541.1
XP_016885030.1
9
1716
XM_017029542.1
XP_016885031.1
8
1563
GenBank mRNA
AF035360.1
-
6
1136
AF041208.1
-
6
1450
AF041210.1
-
5
939
AF230976.1
-
6
1235
AF230977.1
-
6
1235
AF269101.1
-
7
1358
AK025187.1
-
5
421
AK226104.1
-
6
1154
AK291215.1
-
6
1332
AK297205.1
-
7
1246
AK299061.1
-
6
1275
AK315095.1
-
6
1281
AY539962.1
-
5
432
AY539983.1
-
5
475
AY539994.1
-
6
1202
AY539997.1
-
4
349
AY540009.1
-
2
119
AY540010.1
-
2
224
AY540011.1
-
1
181
AY540012.1
-
2
119
AY540013.1
-
1
447
AY540022.1
-
5
1049
AY540023.1
-
5
935
BC053626.1
-
6
1307
6163881
Y13667.1
-
6
1235
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00166057_m1
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0