Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 11 | 1612 | ||
NM_001318368.1 | NP_001305297.1 | 11 | 1612 | |||
XM_005266216.4 | XP_005266273.1 | 11 | 970 | |||
XM_011534864.1 | XP_011533166.1 | 10 | 1533 | |||
XM_011534865.2 | XP_011533167.1 | 7 | 1657 | |||
XM_017020317.1 | XP_016875806.1 | 10 | 968 | |||
XM_017020318.1 | XP_016875807.1 | 7 | 702 | |||
GenBank mRNA | AK296084.1 | - | 7 | 602 | ||
AK296875.1 | - | 10 | 955 | |||
AY702976.1 | - | 10 | 1016 | |||
BC069634.1 | - | 11 | 1097 | 7262421 | ||
BC094784.1 | - | 10 | 1554 | |||
BC104492.1 | - | 11 | 1100 | 40032854 | ||
BC105950.1 | - | 11 | 1100 | 40032852 | ||
BC107601.1 | - | 11 | 1099 | 40032849 | ||
L23208.1 | - | 3 | 210 | |||
X85753.1 | - | 11 | 1100 |
Gene Symbol: ADRA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309989.1 | - | 1333 |