Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324069.1 | NP_001310998.1 | 7 | 622 | ||
NM_001324070.1 | NP_001310999.1 | 3 | 364 | |||
NM_001324071.1 | NP_001311000.1 | 4 | 398 | |||
NM_001324072.1 | NP_001311001.1 | 6 | 399 | |||
NM_001324074.1 | NP_001311003.1 | 5 | 501 | |||
NM_001324075.1 | NP_001311004.1 | 6 | 533 | |||
NM_001324077.1 | NP_001311006.1 | 6 | 401 | |||
NM_001324078.1 | NP_001311007.1 | 5 | 499 | |||
NM_001799.3 | NP_001790.1 | 6 | 535 | |||
NR_136690.1 | 6 | 535 | ||||
XM_011543094.2 | XP_011541396.1 | 3 | 205 | |||
GenBank mRNA | AK026509.1 | - | 6 | 420 | ||
BC000834.2 | - | 6 | 374 | 3455907 | ||
BC005298.1 | - | 6 | 442 | 3997991 | ||
BI520184.1 | - | 6 | 536 | |||
BU660651.1 | - | 6 | 364 | |||
KU177970.1 | - | 6 | 354 | |||
KU177971.1 | - | 5 | 318 | |||
L20320.1 | - | 7 | 529 | |||
X77303.1 | - | 6 | 397 | |||
X77743.1 | - | 6 | 388 | |||
X79193.1 | - | 6 | 444 | |||
Y13120.1 | - | 6 | 457 |