Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324069.1 | NP_001310998.1 | 11 | 1074 | ||
NM_001324070.1 | NP_001310999.1 | 7 | 816 | |||
NM_001324071.1 | NP_001311000.1 | 8 | 850 | |||
NM_001324072.1 | NP_001311001.1 | 10 | 851 | |||
NM_001324074.1 | NP_001311003.1 | 9 | 953 | |||
NM_001324075.1 | NP_001311004.1 | 10 | 985 | |||
NM_001324077.1 | NP_001311006.1 | 10 | 853 | |||
NM_001324078.1 | NP_001311007.1 | 9 | 951 | |||
NM_001799.3 | NP_001790.1 | 10 | 987 | |||
NR_136690.1 | 9 | 868 | ||||
XM_011543094.2 | XP_011541396.1 | 7 | 657 | |||
GenBank mRNA | AK026509.1 | - | 10 | 872 | ||
BC000834.2 | - | 10 | 826 | 3455907 | ||
BC005298.1 | - | 10 | 894 | 3997991 | ||
KU177970.1 | - | 10 | 806 | |||
KU177971.1 | - | 9 | 770 | |||
L20320.1 | - | 11 | 981 | |||
X77303.1 | - | 10 | 849 | |||
X77743.1 | - | 10 | 840 | |||
X79193.1 | - | 10 | 896 | |||
Y13120.1 | - | 10 | 909 |