Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMYA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153610.4 | NP_705838.3 | 2 | 9294 | ||
XM_017009212.1 | XP_016864701.1 | 2 | 9162 | |||
GenBank mRNA | AK123466.1 | - | 1 | 652 | ||
AL831966.1 | - | 1 | 1427 | |||
AL832368.1 | - | 1 | 901 | |||
AL834252.2 | - | 1 | 1011 | |||
BC111529.2 | - | 1 | 604 | |||
BC111530.2 | - | 1 | 604 | |||
BX647628.1 | - | 1 | 998 | |||
BX647646.1 | - | 1 | 998 |