Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMYA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153610.4 | NP_705838.3 | 2 | 10547 | ||
XM_017009212.1 | XP_016864701.1 | 2 | 10415 | |||
GenBank mRNA | AF533705.1 | - | 1 | 1141 | ||
AK123466.1 | - | 1 | 1905 | |||
AL831966.1 | - | 1 | 2680 | |||
AL831986.1 | - | 1 | 1099 | |||
AL832021.1 | - | 1 | 2921 | |||
AL832368.1 | - | 1 | 2154 | |||
AL834252.2 | - | 1 | 2264 | |||
BC111529.2 | - | 1 | 1857 | |||
BC111530.2 | - | 1 | 1857 | |||
BX647628.1 | - | 1 | 2251 | |||
BX647646.1 | - | 1 | 2251 |