Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198834.2 | NP_942131.1 | 4 | 892 | ||
NM_198836.2 | NP_942133.1 | 4 | 460 | |||
NM_198837.1 | NP_942134.1 | 2 | 530 | |||
NM_198838.1 | NP_942135.1 | 3 | 641 | |||
NM_198839.2 | NP_942136.1 | 8 | 1566 | |||
XM_005257267.4 | XP_005257324.1 | 4 | 373 | |||
XM_006721853.1 | XP_006721916.1 | 4 | 903 | |||
XM_011524701.1 | XP_011523003.1 | 4 | 813 | |||
XM_011524703.1 | XP_011523005.1 | 4 | 445 | |||
XM_011524704.2 | XP_011523006.1 | 4 | 378 | |||
XM_017024553.1 | XP_016880042.1 | 5 | 927 | |||
XM_017024554.1 | XP_016880043.1 | 5 | 927 | |||
XM_017024555.1 | XP_016880044.1 | 4 | 526 | |||
GenBank mRNA | AB371587.1 | - | 4 | 439 | ||
AJ534888.1 | - | 2 | 213 | |||
AJ534889.1 | - | 3 | 277 | |||
AK308905.1 | - | 5 | 865 | |||
AK309084.1 | - | 4 | 448 | |||
AY237919.1 | - | 2 | 269 | |||
AY315619.1 | - | 3 | 641 | |||
AY315620.1 | - | 2 | 530 | |||
AY315621.1 | - | 4 | 460 | |||
AY315622.1 | - | 5 | 571 | |||
AY315623.1 | - | 8 | 1536 | |||
AY315625.1 | - | 4 | 862 | |||
AY315626.1 | - | 5 | 973 | |||
AY315627.1 | - | 2 | 269 | |||
BC137287.1 | - | 4 | 397 | 9021284 |