Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082533.1 | NP_001076002.1 | 3 | 1196 | ||
NM_001082534.1 | NP_001076003.1 | 3 | 761 | |||
NM_020178.4 | NP_064563.1 | 2 | 1070 | |||
GenBank mRNA | AB001597.1 | - | 2 | 84 | ||
AB036836.1 | - | 3 | 986 | |||
AF288385.1 | - | 3 | 752 | |||
AK313001.1 | - | 2 | 646 | |||
AL161812.1 | - | 2 | 86 | |||
AY358509.1 | - | 2 | 364 | |||
BC020577.1 | - | 3 | 728 | 4181377 | ||
BC029865.1 | - | 3 | 730 | |||
BC047456.1 | - | 2 | 983 | 5276935 | ||
BC068462.1 | - | 2 | 1029 | 5260321 | ||
CR749607.1 | - | 3 | 532 |