Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008539.3 | NP_001008539.3 | 9 | 1503 | ||
NM_001164771.1 | NP_001158243.1 | 9 | 1499 | |||
NM_003046.5 | NP_003037.4 | 9 | 1496 | |||
XM_005273610.4 | XP_005273667.1 | 10 | 1564 | |||
XM_005273611.4 | XP_005273668.1 | 10 | 1467 | |||
XM_005273612.4 | XP_005273669.1 | 10 | 1561 | |||
XM_017013746.1 | XP_016869235.1 | 9 | 1502 | |||
XM_017013747.1 | XP_016869236.1 | 10 | 1561 | |||
GenBank mRNA | AK314287.1 | - | 9 | 1501 | ||
AL832016.2 | - | 4 | 392 | |||
BC069648.1 | - | 9 | 1406 | 7262473 | ||
BC104905.1 | - | 9 | 1389 | 8143908 | ||
BC113661.1 | - | 9 | 1389 | 8322713 | ||
BC143583.1 | - | 9 | 1392 | 9052094 | ||
D29990.1 | - | 8 | 1331 | |||
U76368.1 | - | 9 | 1522 | |||
U76369.1 | - | 4 | 325 |