Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000809.3 | NP_000800.2 | 6 | 1680 | ||
NM_001204266.1 | NP_001191195.1 | 6 | 710 | |||
NM_001204267.1 | NP_001191196.1 | 6 | 710 | |||
GenBank mRNA | AK090780.1 | - | 6 | 1680 | ||
BC035055.1 | - | 6 | 809 | 5259426 | ||
CD013892.1 | - | 6 | 714 | |||
CD013893.1 | - | 6 | 714 | |||
KJ534840.1 | - | 6 | 697 | |||
U30461.1 | - | 6 | 711 |