Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000390.3 | NP_000381.1 | 14 | 1723 | ||
NM_001320959.1 | NP_001307888.1 | 14 | 1720 | |||
XM_017029242.1 | XP_016884731.1 | 14 | 1719 | |||
XM_017029243.1 | XP_016884732.1 | 14 | 1810 | |||
XM_017029244.1 | XP_016884733.1 | 14 | 1708 | |||
XM_017029245.1 | XP_016884734.1 | 14 | 1810 | |||
XM_017029246.1 | XP_016884735.1 | 14 | 1689 | |||
GenBank mRNA | AK291160.1 | - | 14 | 1712 | ||
AK299323.1 | - | 11 | 1428 | |||
AK312443.1 | - | 14 | 1712 | |||
M83773.1 | - | 8 | 1039 | |||
X57637.1 | - | 7 | 805 | |||
X78121.1 | - | 14 | 1720 |