Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009939.2 | NP_001009939.1 | 2 | 462 | ||
NM_002688.5 | NP_002679.2 | 3 | 257 | |||
GenBank mRNA | AK056273.1 | - | 2 | 439 | ||
AK295940.1 | - | 3 | 126 | |||
BC025261.1 | - | 3 | 221 | 4622405 | ||
BI601927.1 | - | 2 | 467 | |||
CR456545.1 | - | 3 | 137 | |||
KJ534756.1 | - | 3 | 132 | |||
KJ534757.1 | - | 3 | 132 | |||
U74628.1 | - | 3 | 219 | |||
U85431.1 | - | 3 | 197 | |||
Y11593.1 | - | 3 | 177 |
Gene Symbol: SEPT5-GP1BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037611.1 | 2 | 1677 | |||
NR_037612.1 | 2 | 181 | ||||
GenBank mRNA | AB209168.1 | - | 2 | 1677 | ||
U59632.1 | - | 2 | 181 |